Learn More
Meso Scale Discovery GAPDH Kit
SDP
Catalog No. 5000078788
Encompass
The Meso Scale Discovery GAPDH Kit is a 96-well plate assay for the quantitative measurement of GAPDH in cell and tissue lysates It contains a 96-well plate coated with a GAPDH capture antibody a detection antibody conjugated to an electrochemiluminescent label and calibrator containing recombinant GAPDH The assay has a detection range of 1 56-200 ng/mL and takes 3 5 hours to complete It utilizes MSD s electrochemiluminescence technology for high sensitivity and wide dynamic range The kit includes all necessary reagents and can process up to 40 samples in duplicate It is designed for research use only
Catalog No. 50-000-78788 Supplier Meso Scale Discovery Supplier No. K151PWD1
The Meso Scale Discovery GAPDH Kit is a 96-well plate assay for the quantitative measurement of GAPDH in cell and tissue lysates It contains a 96-well plate coated with a GAPDH capture antibody a detection antibody conjugated to an electrochemiluminescent label and calibrator containing recombinant GAPDH The assay has a detection range of 1 56-200 ng/mL and takes 3 5 hours to complete It utilizes MSD s electrochemiluminescence technology for high sensitivity and wide dynamic range The kit includes all necessary reagents and can process up to 40 samples in duplicate It is designed for research use only