Learn More
Meso Scale Discovery GAPDH Kit
SDP
Catalog No. 5000081666
Encompass
The Meso Scale Discovery GAPDH Kit (K151PWD) is a 96-well plate assay kit for quantifying GAPDH protein in cell and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody Read Buffer Sample Diluent and lyophilized GAPDH protein standard The electrochemiluminescent assay has a typical sensitivity of 1-2 pg/mL and a broad dynamic range up to 4 logs The kit can be used to normalize target protein levels or as a reporter gene assay It is suitable for cells and tissues including human mouse rat monkey canine porcine and rabbit
Catalog No. 50-000-81666 Supplier Meso Scale Discovery Supplier No. K151PWD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery GAPDH Kit (K151PWD) is a 96-well plate assay kit for quantifying GAPDH protein in cell and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody Read Buffer Sample Diluent and lyophilized GAPDH protein standard The electrochemiluminescent assay has a typical sensitivity of 1-2 pg/mL and a broad dynamic range up to 4 logs The kit can be used to normalize target protein levels or as a reporter gene assay It is suitable for cells and tissues including human mouse rat monkey canine porcine and rabbit