Learn More
Enzo Life Sciences GAPDH monoclonal antibody (GA1R) (biotin conjugate) (50µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2770050
Encompass_Preferred
Clone: GA1R. Isotype: IgG1. Immunogen: Full-length recombinant GAPDH. Species reactivity: Human, Mouse, Rat, Yeast, Rabbit, Insect, Hamster, E. coli, Chicken. Specificity: Recognizes GAPDH in multiple species, including human, mouse, rat, bacteria, chicken, hamster, insect, rabbit and yeast. Applications: ELISA, Flow Cytometry, ICC, IF, IHC (PS), WB. Purity: Purified. Formulation: Liquid. In PBS containing a proprietary stabilizer and 0.02% sodium azide.
Catalog No. 50-201-2821