Learn More
Biomatik Corporation Gapdh Monoclonal Antibody
Supplier:  Biomatik Corporation CAC11883
Encompass_Preferred
Gapdh Monoclonal Antibody is a Monoclonal Antibody to GAPDH. The Gapdh Monoclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP, IF, FC. Host: Mouse. Immunogen: Recombinant Human Gapdh Protein (3-335Aa). Applications: ELISA, WB, IHC, IP, IF, FC. Target Synonyms: GAPDH. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7607