Learn More
Medchemexpress LLC GAPDH Mouse His 10ug
SDP
Catalog No. 5000179892
Encompass_Preferred
The GAPDH protein has dual roles in glycolysis and nuclear function It also affects the organization and assembly of the cytoskeleton and promotes the association of microtubules and membranes GAPDH Protein Mouse (His) is the recombinant mouse-derived GAPDH protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-79892 Supplier Medchemexpress LLC Supplier No. HYP7412910UG
The GAPDH protein has dual roles in glycolysis and nuclear function It also affects the organization and assembly of the cytoskeleton and promotes the association of microtubules and membranes GAPDH Protein Mouse (His) is the recombinant mouse-derived GAPDH protein expressed by E coli with N-6 His labeled tag