Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GPR35 Antibody Blocking Peptide
SDP

Catalog No. NBP224640SP
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224640SP 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224640SP Supplier Novus Biologicals™ Supplier No. NBP224640PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Blocking peptide for NBP2-24640

Application Notes:This peptide is useful as a blocking peptide for NBP2-24640.

Specifications

Gene ID (Entrez) 2859
Purity Protein A purified
Common Name GPR35
Concentration 0.5 mg/mL
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation PBS containing 0.05% BSA
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP2-24640. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias G protein-coupled receptor 35, G-protein coupled receptor 35, KYNA receptor, Kynurenic acid receptor
Gene Symbol GPR35
Quantity 0.05 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.