Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gapdh, rabbit muscle | 9001-50-7 | 73,600 | 500 U
SDP

Catalog No. 5000396785
Encompass_Preferred

Glyceraldehyde phosphate dehydrogenase (EC 1.2.1.12) is an enzyme that targets anti-thymocyte and anti-apoptotic agents. It catalyzes the chain oxidation of reduced nicotinamide adenine dinucleotide by perhydroxyl radicals. This recombinant product is suitable for enzymatic determination of glycerol and triglycerides when combined with glycerol kinase.

  • Solid appearance, white to light yellow
  • pH stable between 6.5 and 10.0, with an optimum pH of 9.0
  • Soluble in H₂O at concentrations ≥ 25 mg/mL
  • For research use only

Catalog No. 50-003-96785 Supplier Medchemexpress LLC Supplier No. HYP2804500U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glyceraldehyde phosphate dehydrogenase (EC 1.2.1.12) is an enzyme that targets anti-thymocyte and anti-apoptotic agents. It catalyzes the chain oxidation of reduced nicotinamide adenine dinucleotide by perhydroxyl radicals. This recombinant product is suitable for enzymatic determination of glycerol and triglycerides when combined with glycerol kinase.

  • Solid appearance, white to light yellow
  • pH stable between 6.5 and 10.0, with an optimum pH of 9.0
  • Soluble in H₂O at concentrations ≥ 25 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.