Learn More
Medchemexpress LLC GAPDH, rabbit muscle | 9001-50-7 | ≥75 U/mg protein | about 73,600 | 100 U
SDP
Catalog No. 5000399474
Encompass_Preferred
Glyceraldehyde phosphate dehydrogenase (EC 1.2.1.12) is the target of anti-thymocyte and anti-apoptotic agents. It catalyzes the chain oxidation of reduced nicotinamide adenine dinucleotide by perhydroxyl radicals. This product is recombinantly produced and can be used for enzymatic determination of glycerol and triglycerides after combining with glycerol kinase.
  • Catalyzes the chain oxidation of reduced nicotinamide adenine dinucleotide by perhydroxyl radicals.
  • Recombinantly produced.
  • Can be used for enzymatic determination of glycerol and triglycerides.
Catalog No. 50-003-99474 Supplier Medchemexpress LLC Supplier No. HYP2804100U
Glyceraldehyde phosphate dehydrogenase (EC 1.2.1.12) is the target of anti-thymocyte and anti-apoptotic agents. It catalyzes the chain oxidation of reduced nicotinamide adenine dinucleotide by perhydroxyl radicals. This product is recombinantly produced and can be used for enzymatic determination of glycerol and triglycerides after combining with glycerol kinase.
  • Catalyzes the chain oxidation of reduced nicotinamide adenine dinucleotide by perhydroxyl radicals.
  • Recombinantly produced.
  • Can be used for enzymatic determination of glycerol and triglycerides.