Learn More
Santa Cruz Biotechnology Anti-GAPDH Antibody (0411), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC47724
Encompass
GAPDH Antibody (0411) is recommended for detection of GAPDH of human origin by WB, IP, IF and IHC(P); not recommended for detection of GAPDH of mouse or rat origin.
Catalog No. NC9666051