Learn More
Cayman Chemical GAPDHC-Term RabbIt Monclo 50uL
SDP
Catalog No. 502947198
Encompass_Preferred
A synthetic peptide corresponding to an integrin cell adhesion sequence a component of radioligands and peptidomimetics that have been used in the study of integrin expression and activity in vivo
Catalog No. 50-294-7198 Supplier Cayman Chemical Supplier No. 3237850 UL
A synthetic peptide corresponding to an integrin cell adhesion sequence a component of radioligands and peptidomimetics that have been used in the study of integrin expression and activity in vivo