Learn More
Aviva Systems Biology Corp GAPDHS Antibody - N-terminal region (ARP40187_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40187_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAPDHS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GAPDHS. Alternate names: GAPD2, GAPDH-2, GAPDS, HSD-35ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57328