Learn More
Santa Cruz Biotechnology Anti-GAPDH Antibody (G-9), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC365062
Encompass
Anti-GAPDH Antibody (G-9) is recommended for detection of GAPDH and GAPDH-2 of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA.
Catalog No. NC0888776