Learn More
Medchemexpress LLC HY-107569 1mg Medchemexpress, Garcinol CAS:78824-30-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1075691MG
Encompass_Preferred
Medchemexpress, HY-107569 1mg Garcinol CAS:78824-30-3 Garcinol, a polyisoprenylated benzophenone harvested from Garcinia indica, exerts anti-cholinesterase properties towards acetyl cholinesterase (AChE) and butyrylcholinesterase (BChE) with IC50s of 0.66 µM and 7.39 µM, respectively. Garcinol also inhibits histone acetyltransferases (HATs, IC50= 7 μM) and p300/CPB-associated factor (PCAF, IC50 = 5 μM). Garcinol has anti-inflammatory and anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8433