Learn More
Active Motif Garcinol
SDP
Catalog No. NC1840892
Encompass
Active Motif's Garcinol is a potent inhibitor of histone acetyltransferases p300 (IC50 = 7 M) and pCAF (IC50 = 5 M). It induces apoptosis in various cell lines and promotes expansion of human hematopoietic stem cells. It is cell permeable.
Catalog No. NC1840892 Supplier Active Motif Supplier No. 14076
Active Motif's Garcinol is a potent inhibitor of histone acetyltransferases p300 (IC50 = 7 M) and pCAF (IC50 = 5 M). It induces apoptosis in various cell lines and promotes expansion of human hematopoietic stem cells. It is cell permeable.