Learn More
Abcam Garcinol, HAT p300 and PCAF inhibitor, 25MG
Catalog No. 502706559
Encompass_Preferred
MW 602.8 Da, Purity >95%. Potent histone acetyltransferase p300 and PCAF inhibitor (IC₅₀ values are 7 and 5 μM for p300 and PCAF respectively). Pleiotropic agent. Strongly inhibits chromatin transcription and induces apoptosis. Inhibits histone acetylation in vivo. Antioxidant and chemoprotective agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6559 Supplier Abcam Supplier No. AB14150325MG
MW 602.8 Da, Purity >95%. Potent histone acetyltransferase p300 and PCAF inhibitor (IC₅₀ values are 7 and 5 μM for p300 and PCAF respectively). Pleiotropic agent. Strongly inhibits chromatin transcription and induces apoptosis. Inhibits histone acetylation in vivo. Antioxidant and chemoprotective agent.
The product is subject to the following: Abcam Restricted Use Statement