Learn More
Medchemexpress LLC Garcinone D | 107390-08-9 | 10 MM 1 ML
SDP
Catalog No. 5000419173
Encompass_Preferred
Garcinone D is an activator of the STAT3/Cyclin D1 and Nrf2/HO-1 pathways, and an inhibitor of CDK2/CyclinE1 (IC50 for CDK2/CyclinE1 is 28.23 μM). It promotes neural stem cell proliferation by activating STAT3 phosphorylation and Cyclin D1 expression, and enhancing the Nrf2/HO-1 signaling pathway. Additionally, it blocks the tumor cell cycle by inhibiting CDK2/CyclinE1 and can inhibit prostate and breast cancer.
  • Activator of STAT3/Cyclin D1 and Nrf2/HO-1 pathways
  • Inhibitor of CDK2/CyclinE1
  • Promotes neural stem cell proliferation
  • Blocks tumor cell cycle
  • Regulates C17.2 neural stem cell viability
  • Promotes G1-S phase transition and arrests cell cycle in S phase in C17.2 neural stem cells
  • Increases p-STAT3, Cyclin D1, Nrf2, and HO-1 proteins in C17.2 neural stem cells
Catalog No. 50-004-19173 Supplier Medchemexpress LLC Supplier No. HYN695310MM/1ML
Garcinone D is an activator of the STAT3/Cyclin D1 and Nrf2/HO-1 pathways, and an inhibitor of CDK2/CyclinE1 (IC50 for CDK2/CyclinE1 is 28.23 μM). It promotes neural stem cell proliferation by activating STAT3 phosphorylation and Cyclin D1 expression, and enhancing the Nrf2/HO-1 signaling pathway. Additionally, it blocks the tumor cell cycle by inhibiting CDK2/CyclinE1 and can inhibit prostate and breast cancer.
  • Activator of STAT3/Cyclin D1 and Nrf2/HO-1 pathways
  • Inhibitor of CDK2/CyclinE1
  • Promotes neural stem cell proliferation
  • Blocks tumor cell cycle
  • Regulates C17.2 neural stem cell viability
  • Promotes G1-S phase transition and arrests cell cycle in S phase in C17.2 neural stem cells
  • Increases p-STAT3, Cyclin D1, Nrf2, and HO-1 proteins in C17.2 neural stem cells