Learn More
TARGETMOL CHEMICALS INC Garcinone D 1MG
SDP
Catalog No. 502875836
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Garcinone D shows significant cytotoxicity against the CEM-SS cell line, with IC(50) value of 3.2 microg/ml; it exhibits dose-dependent enzyme-based microsomal aromatase inhibitory activity. Garcinone D inhibits p65 activation with IC50 values of 3.2 microM. Purity 99.94%
Catalog No. 50-287-5836 Supplier TARGETMOL CHEMICALS INC Supplier No. TN16741MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Garcinone D shows significant cytotoxicity against the CEM-SS cell line, with IC(50) value of 3.2 microg/ml; it exhibits dose-dependent enzyme-based microsomal aromatase inhibitory activity. Garcinone D inhibits p65 activation with IC50 values of 3.2 microM. Purity 99.94%