Learn More
Medchemexpress LLC Garcinone E | 112649-21-5 | MFCD12756397 | 98.2% | 464.55 g/mol | C28H32O6 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1239011MG
Encompass_Preferred
Garcinone E is a natural xanthone compound isolated from the rind of Garcinia mangostana (mangosteen). It is reported to induce apoptosis and inhibit cancer cell migration, and is used as a research reagent in cellular and biochemical studies of anticancer activity.
  • Natural xanthone compound.
  • Induces apoptosis and inhibits cancer cell migration.
  • Suitable for in vitro cellular and biochemical assays.
  • High purity, approximately 98.2%.
  • Available in small research quantities for assay development and mechanistic studies.
Catalog No. 50-002-36811