Learn More
Description
Includes
- 15 tests each for nitrogen, phosphorus and potassium; 30 tests for soil pH (range 4.0 to 9.0)
- Diagrammed instructions
- Laminated color charts
- Garden Guide manual
- LaMotte™ Soil Handbook
- A Study Of Soil Science (downloadable)
Measures nitrogen, phosphorus, potassium and soil pH.
- New hinged carrying case with foam liner hold all components securely
- Includes the LaMotte Soil Handbook, Garden Guide Manual, instructions, and free downloadable resource, A Study of Soil Science handbook
- Grades 5+
Specifications
Specifications
| Grade | 5 |
| For Use With (Application) | Soil science education or garden analysis by measuring nitrogen, phosphorus, potassium and soil ph |
| Includes | 120mL/250mL pH indicator, Phosphorus and Potassium extracting solution and Nitrogen extracting solution, 10g/30g Nitrogen indicator powder, 15mL/15mL Phosphorus indicator reagent, 30/50 Phosphorus and Potassium indicator tablets, 15mL/2 x 15mL Potassium test solution, 1 to 8 mL Plastic test tubes with caps, 1/1 test tube brush, 3/3 Plastic transfer pipets, 1/1 0.25g and 0.5g Plastic spoon, 1/1 Nitrogen, Phosphorus, wide range pH and Potassium color chart, manual, study of soil science, 1/1 LaMotte™ soil handbook |
| Class Size | 1 Class |
| Product Type | Garden Guide Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

