Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Garden Guide Kit
SDPSCI_ED

Catalog No. S85039R Shop All LaMotte Company Products
Change view
Click to view available options
Product Type:
Garden Guide Kit
Reagent Refill
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S85039R Reagent Refill
S85039 Garden Guide Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S85039R Supplier LaMotte™ Supplier No. R5679
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Reagent Refill

Convenient kit for classroom soil testing and garden analysis.

Measures nitrogen, phosphorus, potassium and soil pH.

  • New hinged carrying case with foam liner hold all components securely
  • Includes the LaMotte Soil Handbook, Garden Guide Manual, instructions, and free downloadable resource, A Study of Soil Science handbook
  • Grades 5+

Specifications

Grade 5
For Use With (Application) Soil science education or garden analysis by measuring nitrogen, phosphorus, potassium and soil ph
Class Size 1 Class
Product Type Reagent Refill
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.