Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte Garden Guide Soil Test Kit
SDPSCI_ED

Catalog No. S89510 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
5 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89510 5 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89510 Supplier LaMotte Supplier No. 567902
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Garden Guide Soil Test Kit is ideal for checking soil nutrient levels in school gardens and landscapes.

  • Measure nitrogen, phosphorus, potassium (15 tests each) and soil pH (30 tests)
  • Compare results to a laminated color chart
  • Diagrammed instructions
  • All hardware supplies' Garden Guid Manual, LaMotte Soil Handbook and free on-line resource A Study of Soil Science.

Specifications

Grade 5 to 15
Age 11 to 20
For Use With (Application) Checking Soil Nutrient Levels
Includes 15 Nitrogen Tests, 15 Phosphorus Tests, 15 Potassium Tests, 30 pH Tests
Class Size 1 Class
Product Type Soil Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.