Learn More
Grainger Garden Hose Hanger Steel 13 in W Green
Catalog No. 502766260
Encompass_Preferred
The product is a Dayton 16X130 Belt, an industrial V-belt with a top width of 16/32 inches and a length of 130 inches. It features a conventional belt construction with friction rubber on the sides to grip the pulley. The belt is designed for use with sheaves having up to a 7 inch pitch diameter. It can transmit up to 15 horsepower at 1750 feet per minute. The belt is manufactured in the USA by Dayco with a quality rubber compound to provide flexibility and resistance to heat, oils, and chemicals. It meets RMA standards for static conductivity. The belt is sold individually and available for fast shipping from Grainger.
Catalog No. 50-276-6260 Supplier Grainger Supplier No. 16X130
The product is a Dayton 16X130 Belt, an industrial V-belt with a top width of 16/32 inches and a length of 130 inches. It features a conventional belt construction with friction rubber on the sides to grip the pulley. The belt is designed for use with sheaves having up to a 7 inch pitch diameter. It can transmit up to 15 horsepower at 1750 feet per minute. The belt is manufactured in the USA by Dayco with a quality rubber compound to provide flexibility and resistance to heat, oils, and chemicals. It meets RMA standards for static conductivity. The belt is sold individually and available for fast shipping from Grainger.