Learn More
Medchemexpress LLC HY-N6675 50mg Medchemexpress, Gardenia yellow CAS:94238-00-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN667550MG
Encompass_Preferred
Medchemexpress, HY-N6675 50mg Gardenia yellow CAS:94238-00-3 Gardenia yellow is an active member of crocin, increases mRNA expression of SIRT3, and acts as an orally active antidepressant agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1565