Learn More
TARGETMOL CHEMICALS INC Gardenin A 1MG
SDP
Catalog No. 502875829
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Gardenin A shows neurotrophic effects via activating MAPK/ERK, PKC, and PKA. Gardenin A exhibits antidepressant, anticonvulsant and anxiolytic effects. Purity 95.15%
Catalog No. 50-287-5829 Supplier TARGETMOL CHEMICALS INC Supplier No. TN41141MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Gardenin A shows neurotrophic effects via activating MAPK/ERK, PKC, and PKA. Gardenin A exhibits antidepressant, anticonvulsant and anxiolytic effects. Purity 95.15%