Learn More
Cayman Chemical Gardenoside, Purity: ≥95%, CAS: 24512-62-7, 10 mg
SDP
Catalog No. 502947971
Encompass_Preferred
An iridoid glycoside with anti-inflammatory and antinociceptive activities; decreases free fatty acid-induced accumulation of lipid droplets and increases in TNF-α, IL-6, and IL-1β levels in HepG2 cells at 10 and 20 µM; reduces increases in the expression of purinergic P2X3 and P2X7 receptors, as well as reverses decreases in mechanical withdrawal thresholds in a rat model of chronic constriction injury-induced neuropathic pain
Catalog No. 50-294-7971 Supplier Cayman Chemical Supplier No. 3438110 MG
An iridoid glycoside with anti-inflammatory and antinociceptive activities; decreases free fatty acid-induced accumulation of lipid droplets and increases in TNF-α, IL-6, and IL-1β levels in HepG2 cells at 10 and 20 µM; reduces increases in the expression of purinergic P2X3 and P2X7 receptors, as well as reverses decreases in mechanical withdrawal thresholds in a rat model of chronic constriction injury-induced neuropathic pain