Learn More
Enzo Life Sciences Gardiquimod (100mg)
SDP
Supplier:  Enzo Life Sciences ALX420040M100
Encompass_Preferred
Selective ligand for human or mouse Toll-like receptor 7 (TLR7). Induces the activation of NF-κB in HEK 293 cells expressing TLR7. At high concentrations (3µg/ml) slightly activates TLR8. More active than Imiquimod (ALX-420-039). Alternative name: 1-(4-Amino-2-ethylaminomethylimidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol. Purity: ≥98% (HPLC (UV)). Appearance: White to off-white solid. Solubility: Soluble in DMSO (>40mg/ml), dioxane (7mg/ml), methanol (>40mg/ml) or acetonitrile (2mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9118