Learn More
Cayman Chemical GardIquImod 10mg
SDP
Catalog No. 502985144
Encompass_Preferred
A selective agonist of human TLR7; enhances the immunosuppressive activity of Treg cells at 0.5 to 2 UG/ml
Catalog No. 50-298-5144 Supplier Cayman Chemical Supplier No. 1887710 MG
A selective agonist of human TLR7; enhances the immunosuppressive activity of Treg cells at 0.5 to 2 UG/ml