Learn More
Abcam Gardiquimod, toll-like receptor 7 (TLR7) agonist, 5MG
Catalog No. 502706575
Encompass_Preferred
MW 313.4 Da, Purity >98%. Selective TLR7 agonist. Inhibits HIV-1 reverse transcriptase. Induces pro-inflammatory cytokine production in thymocytes. Antiproliferative effects. Shows immunomodulatory effects in vivo. More active than Imiquimod (ab141410).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6575 Supplier Abcam Supplier No. AB1414855MG
MW 313.4 Da, Purity >98%. Selective TLR7 agonist. Inhibits HIV-1 reverse transcriptase. Induces pro-inflammatory cytokine production in thymocytes. Antiproliferative effects. Shows immunomodulatory effects in vivo. More active than Imiquimod (ab141410).
The product is subject to the following: Abcam Restricted Use Statement