Learn More
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| CAS Min % | 76.0, 14.98, 3.6, 3.32 |
| CAS Max % | 79.0, 15.59, 3.75, 3.46 |
| Physical Form | Liquid |
| Color | Yellow |
| pH | Acidic |
| For Use With (Equipment) | Water Analyzer |
| Solubility | Miscible with water |
| Solution Type | Calibration Solution |
| Type | Color Reference Standard |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
