Learn More
ATCC Gardnerella vaginalis
Catalog No. 502382430
Encompass
Gardnerella vaginalis strain AmMS 117 is a whole-genome sequenced clinical isolate that can be used as a quality control strain for MicroScan products.
Catalog No. 50-238-2430 Supplier ATCC Supplier No. 49145
Gardnerella vaginalis strain AmMS 117 is a whole-genome sequenced clinical isolate that can be used as a quality control strain for MicroScan products.