Learn More
MSC Chemetall Manganese Phosphating Process, Low Temperature, Immersion, Iron & Steel
Catalog No. 502838343
Encompass
GARDOBOND G 4040 is a low temperature manganese phosphating process for iron and steel, applied by immersion. It is designed to enhance wear resistance and reduce friction in sliding parts. This process complies with military and commercial specifications, specifically MIL-DTL-16232G Type M.
  • Enhances wear resistance of sliding parts.
  • Reduces friction in sliding parts.
  • Low temperature manganese phosphating process.
  • Applied by immersion.
  • Produces manganese phosphate coatings with a weight range of 400-2000 milligrams per square foot.
  • Meets military Detail Specification MIL-DTL-16232G Type M.
  • Can reduce processing time, sludge, and scale with an optional accelerator.
Catalog No. 50-283-8343 Supplier MSC Supplier No. 37649704
GARDOBOND G 4040 is a low temperature manganese phosphating process for iron and steel, applied by immersion. It is designed to enhance wear resistance and reduce friction in sliding parts. This process complies with military and commercial specifications, specifically MIL-DTL-16232G Type M.
  • Enhances wear resistance of sliding parts.
  • Reduces friction in sliding parts.
  • Low temperature manganese phosphating process.
  • Applied by immersion.
  • Produces manganese phosphate coatings with a weight range of 400-2000 milligrams per square foot.
  • Meets military Detail Specification MIL-DTL-16232G Type M.
  • Can reduce processing time, sludge, and scale with an optional accelerator.