Learn More
MSC Chemetall Gardotest Sol 100, 0.01 M Disodium EDTA, 4 L
Catalog No. 502816164
Encompass
Gardotest Sol 100 Disodium EDTA is a 0.01 M Disodium EDTA (Triplex III) solution specifically formulated for accurate water hardness testing. This reagent is supplied in a convenient 4 L volume for laboratory use.
  • Used for hardness testing applications
  • Solution-based test method
  • Does not contain mercury
  • Not marked as hazardous
  • Provided in 4 L volume
Catalog No. 50-281-6164 Supplier MSC Supplier No. 09848573
Gardotest Sol 100 Disodium EDTA is a 0.01 M Disodium EDTA (Triplex III) solution specifically formulated for accurate water hardness testing. This reagent is supplied in a convenient 4 L volume for laboratory use.
  • Used for hardness testing applications
  • Solution-based test method
  • Does not contain mercury
  • Not marked as hazardous
  • Provided in 4 L volume