Learn More
MSC GardTec Power Cord, NEMA 6-15P, 230V, 72 in, 1 ea
Catalog No. 5000506508
Encompass
This heavy-duty fan power cord is designed for 230V applications, suitable for cabinet cooling fan kits. It features a 45-degree plug and an integrated ground wire, ensuring reliable power connectivity for cooling fans.
  • Heavy duty construction
  • Features a 45 degree plug for convenient access
  • SPT1 wire, 105 C, 300 volts, black
  • NEMA 6-15P plug with #8 ring terminal
  • Provides 72 inch (1829 mm) length
  • UL listed (E189621, E156798)
  • Manufactured in USA
Catalog No. 50-005-06508 Supplier MSC Supplier No. 44906139
This heavy-duty fan power cord is designed for 230V applications, suitable for cabinet cooling fan kits. It features a 45-degree plug and an integrated ground wire, ensuring reliable power connectivity for cooling fans.
  • Heavy duty construction
  • Features a 45 degree plug for convenient access
  • SPT1 wire, 105 C, 300 volts, black
  • NEMA 6-15P plug with #8 ring terminal
  • Provides 72 inch (1829 mm) length
  • UL listed (E189621, E156798)
  • Manufactured in USA