Learn More
Cayman Chemical Garenoxacin, Purity: ≥98%, CAS: 194804-75-6, 10 mg
SDP
Catalog No. 502950089
Encompass_Preferred
A quinolone antibiotic; active against a variety of Gram-positive and Gram-negative bacteria (MIC90s = 0.025-6.25 and 0.0125-100 µg/ml, respectively); inhibits E. coli DNA gyrase supercoiling and S. aureus topoisomerase IV decatenation activities (IC50s = 0.17 and 2.19 mg/L, respectively); selective for S. aureus topoisomerase IV over human topoisomerase II (IC50 = 509.7 µg/L); efficacious against systemic quinolone-resistant S. aureus infection and pneumonia induced by the penicillin-resistant S. pneumoniae clinical isolate D-979 in mice (ED50s = 0.0189 and 0.0278 mg/animal, respectively)
Catalog No. 50-295-0089 Supplier Cayman Chemical Supplier No. 3359910 MG
A quinolone antibiotic; active against a variety of Gram-positive and Gram-negative bacteria (MIC90s = 0.025-6.25 and 0.0125-100 µg/ml, respectively); inhibits E. coli DNA gyrase supercoiling and S. aureus topoisomerase IV decatenation activities (IC50s = 0.17 and 2.19 mg/L, respectively); selective for S. aureus topoisomerase IV over human topoisomerase II (IC50 = 509.7 µg/L); efficacious against systemic quinolone-resistant S. aureus infection and pneumonia induced by the penicillin-resistant S. pneumoniae clinical isolate D-979 in mice (ED50s = 0.0189 and 0.0278 mg/animal, respectively)