Learn More
TARGETMOL CHEMICALS INC Garenoxacin mesylate hydrate
SDP
Catalog No. 502876852
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Garenoxacin mesylate hydrate (Garenoxacin mesylate [USAN]) with potent antimicrobial activity, against common respiratory pathogens, including resistant strains. Purity 99.9%
Catalog No. 50-287-6852 Supplier TARGETMOL CHEMICALS INC Supplier No. T780410MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Garenoxacin mesylate hydrate (Garenoxacin mesylate [USAN]) with potent antimicrobial activity, against common respiratory pathogens, including resistant strains. Purity 99.9%