Learn More
MSC Bare Ground Solutions Garlic Scentry Concentrate, Half Gallon, Liquid, 1:10 Dilution
Catalog No. 502837545
Encompass
Bare Ground Solutions Garlic Scentry Concentrate is a liquid garlic solution that mixes with water to create a spray designed to repel unwanted animals and insects. This chemical-free, all-natural, and environmentally friendly product offers a humane solution for pest control around homes and gardens.
  • Chemical-free and all-natural formulation
  • Environmentally friendly and humane pest repellent
  • Mixes in a 1:10 ratio with water for easy application
  • Absorbed systemically by plants for root to flower protection
  • Effective against a wide range of pests including deer, rabbits, insects, and birds
  • Non-toxic and EPA 25(b) exempt, safe for use around children and pets
  • Does not alter the taste of fruits or vegetables
Catalog No. 50-283-7545 Supplier MSC Supplier No. 47090790
Bare Ground Solutions Garlic Scentry Concentrate is a liquid garlic solution that mixes with water to create a spray designed to repel unwanted animals and insects. This chemical-free, all-natural, and environmentally friendly product offers a humane solution for pest control around homes and gardens.
  • Chemical-free and all-natural formulation
  • Environmentally friendly and humane pest repellent
  • Mixes in a 1:10 ratio with water for easy application
  • Absorbed systemically by plants for root to flower protection
  • Effective against a wide range of pests including deer, rabbits, insects, and birds
  • Non-toxic and EPA 25(b) exempt, safe for use around children and pets
  • Does not alter the taste of fruits or vegetables