Learn More
BioSpec Products Garnet Sharp Particles, 0.3mm diameter
SDP
Catalog No. NC1125707
Encompass_Preferred
Like Silicon Carbide sharp particles, these abrasive particles have sharp cutting edges that may accelerate lysis of plant and tough animal tissue in a Mini-Beadbeater vial.
Catalog No. NC1125707 Supplier BioSpec Products Supplier No. 11079103GAR
Like Silicon Carbide sharp particles, these abrasive particles have sharp cutting edges that may accelerate lysis of plant and tough animal tissue in a Mini-Beadbeater vial.