Learn More
Medchemexpress LLC GARP Latent TGF beta 100ug | 100ug
SDP
Catalog No. 5000187433
Encompass_Preferred
GARP/latent TGF-b1 complexes are potent inducers of Th17 differentiation in the presence of exogenous IL-6 and inducers of Treg in the presence of IL-2 GARP Latent TGF beta Complex Protein Mouse (Biotinylated HEK293 His-Avi) is a biotinylated recombinant protein (GARP (I18-N628) TGF-b1 (L30-S390)) produced by HEK293 cells with His-Avi tag[1][2]
Catalog No. 50-001-87433 Supplier Medchemexpress LLC Supplier No. HYP77678100UG
GARP/latent TGF-b1 complexes are potent inducers of Th17 differentiation in the presence of exogenous IL-6 and inducers of Treg in the presence of IL-2 GARP Latent TGF beta Complex Protein Mouse (Biotinylated HEK293 His-Avi) is a biotinylated recombinant protein (GARP (I18-N628) TGF-b1 (L30-S390)) produced by HEK293 cells with His-Avi tag[1][2]