Learn More
Medchemexpress LLC GARP Latent TGF beta 20ug | 20ug
SDP
Catalog No. 5000187316
Encompass_Preferred
GARP/latent TGF-b1 complexes are potent inducers of Th17 differentiation in the presence of exogenous IL-6 and inducers of Treg in the presence of IL-2 GARP Latent TGF beta Complex Protein Mouse (HEK293 His) is a biotinylated recombinant protein (GARP (I18-N628) TGF-b1 (L30-S390)) produced by HEK293 cells with His tag[1][2]
Catalog No. 50-001-87316 Supplier Medchemexpress LLC Supplier No. HYP7767720UG
GARP/latent TGF-b1 complexes are potent inducers of Th17 differentiation in the presence of exogenous IL-6 and inducers of Treg in the presence of IL-2 GARP Latent TGF beta Complex Protein Mouse (HEK293 His) is a biotinylated recombinant protein (GARP (I18-N628) TGF-b1 (L30-S390)) produced by HEK293 cells with His tag[1][2]