Learn More
Genscript Corporation GARPLatent TGF beta 1 Complex
Supplier:  Genscript Corporation Z04467100
Encompass
GARP Latent TGF Beta is a complex found on surface of many types of cells In Tregs GARP is involved in TCR-mediated activation of Latent TGF- and thus promoting secretion and activation of TGF- Integrin v 8 on the surface of immune cells and other cells recognizes RGD in LAP resulting in the release of mature TGF- from the TGF- GARP complex
Catalog No. 50-000-57227