Learn More
Aviva Systems Biology Corp GARS Antibody - middle region (ARP58758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GARS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GARS. Alternate names: CMT2D, DSMAV, GlyRS, HMN5, SMAD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62413