Learn More
Medchemexpress LLC HY-N6038 5mg Medchemexpress, Gartanin CAS:33390-42-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN60385MG
Encompass_Preferred
Medchemexpress, HY-N6038 5mg Gartanin CAS:33390-42-0 Gartanin is a natural xanthone of mangosteen, with antioxidant, anti-inflammatory, antifungal, neuroprotective and antineoplastic properties. Gartanin induces cell cycle arrest and autophagy and suppresses migration in human glioma cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0446