Learn More
Cayman Chemical Gartnn 10mg
SDP
Catalog No. 502913660
Encompass_Preferred
A derivative of sorafenib selectively inhibits proliferation of BEL-7402 MGC803 and T24 bladder cancer cells (IC50s 5 77 5 21 and 3 97 UM respectively) over non-cancerous HCV-29 cells (IC50 23 19 UM) induces apoptosis in T24 cells at 2-6 UM induces ferroptosis in T24 cells at 6 UM when incubated for 10 hours induces autophagy at 6 UM when incubated for 8 hours
Catalog No. 50-291-3660 Supplier Cayman Chemical Supplier No. 35631 10MG
A derivative of sorafenib selectively inhibits proliferation of BEL-7402 MGC803 and T24 bladder cancer cells (IC50s 5 77 5 21 and 3 97 UM respectively) over non-cancerous HCV-29 cells (IC50 23 19 UM) induces apoptosis in T24 cells at 2-6 UM induces ferroptosis in T24 cells at 6 UM when incubated for 10 hours induces autophagy at 6 UM when incubated for 8 hours