Learn More
Cole-Parmer Cole-Parmer Disposable In-Line Gas/Liquid Filters; Nylon, 99.99% gas, 2 um liquid, 5/pk
Catalog No. NC0063011 Shop All Cole-Parmer Products
Encompass_Preferred
Filter gases or liquids with these microfiber filters. All models have a borosilicate glass filter membrane. See table below for applications and efficiency ratings. Liquid efficiency ratings are 98% of retention of particles of noted size; gas efficiency ratings are percentage retention of 0.01µm particles. Max differential pressure for all filters is 40 psi (60 psi for 0.01 µm units).
Catalog No. NC0063011 Supplier Cole-Parmer Supplier No. 0290940
Filter gases or liquids with these microfiber filters. All models have a borosilicate glass filter membrane. See table below for applications and efficiency ratings. Liquid efficiency ratings are 98% of retention of particles of noted size; gas efficiency ratings are percentage retention of 0.01µm particles. Max differential pressure for all filters is 40 psi (60 psi for 0.01 µm units).