Learn More
GENERAL BIOPHYSICS LLC Environmental Chamber with Gas Analyzer for Animal Studies
Catalog No. NC3361705
Encompass
Environmental Chamber for Mice with Noble Gas MonitoringVolume: 8 liters (supports 10-12 mice).Features: Gas recirculation system with O₂ and CO₂ concentration monitoring, CO₂ scrubbing, integrated noble gas monitor.
Catalog No. NC3361705 Supplier GENERAL BIOPHYSICS LLC Supplier No. ECGAM
Add to Cart
Add to Cart
Request a Quote
Environmental Chamber for Mice with Noble Gas MonitoringVolume: 8 liters (supports 10-12 mice).Features: Gas recirculation system with O₂ and CO₂ concentration monitoring, CO₂ scrubbing, integrated noble gas monitor.