Learn More
Stoelting™ Gas Anesthesia Mask for Stereotaxic Instruments, Rat
SDP
Catalog No. 10000140 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Rat
Catalog No. Animal
10-000-140 Rat
1 options
Catalog No. 10-000-140 Supplier Stoelting™ Supplier No. 51610
Add to Cart
Add to Cart
Stoelting Gas Anesthesia Mask for Stereotaxic Instruments is designed for delivery of gas aneshesia during stereotaxic surgery.
  • Gas anesthesia mask is installed over the incisor bar, with the cone opening toward the open U.
  • Animal (rat or mouse) is installed by placing its teeth over the incisor bar, and sliding the adaptor forward until the cone is snug about the animal's nose, thereby substituting for the nose clamp.
  • Teeth and cone provide a snug, stable hold
  • Anesthetic gas flows in the tubing connector, past the animal's nose, and out the other side.
  • Spent gas may be routed to recycling, exhaust or gas analysis instruments.
Specifications
For Use With (Application) Neuroscience
Animal Rat