Learn More
Description
Inhalant Anesthesia System for Stereotaxic Surgery with Active Gas Waste Removal inlcudes:
- Tabletop and/or Mobile Anesthesia System Complete with Isoflurane Vaporizer
- Active Gas Scavenger
- Continuous Induction Chamber
Features:
- No flowmeters to adjust. Flip a toggle switch to on to start flow of fresh gas, indicator confirms flow
- 3 stations for anesthesia with preset flow rate for any station (500cc-5 LPM)
- Color coded tubing and connector to help user identify which fresh gas supply tubing goes to which station
- Pressure gauge to let user know when pressure has been released from system for filling of vaporizer.
- Generates a 50 LPM negative flow that can be attenuated to appropriate levels and divided between 3 stations.
- Direct vacuum port for use with Vapor-Vac Hooded Induction Chamber
Adjustable negative flow meters easily activated with toggle switches. - Waste anesthetic gases can be discharged into activated charcoal canister or any non re-circulating hood, vent, downdraft/ back draft system or snorkel.
- Available without negative flow generating pump- can then be used to attenuate active, in-house vacuum source.
- Designed for use with the Posi-vac Non-Rebreathing system, Vapor-Vac hooded induction chamber, Stereotaxic nose cone or SCOOP, 3EX Vaporizer. (not included)
- Higher Throughput
- Intuitive system- easy to operate, precise, safe flow rate for fresh gas
Healthy for subjects- less morbidity, less mortality
Specifications
Specifications
| For Use With (Equipment) | Stereotaxic surgery |
| Includes | Tabletop w/ Induction Chamber |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
