Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Gas Anesthesia System
SDP

Catalog No. 10000940 Shop All Stoelting Co Products
Change view
Click to view available options
Includes:
Tabletop w/ Induction Chamber
Tabletop/Mobile w/Induction Chamber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10-000-940 Tabletop/Mobile w/Induction Chamber
10-000-939 Tabletop w/ Induction Chamber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-940 Supplier Stoelting™ Supplier No. 53941
Add to Cart
Add to Cart
Request a Quote

Stoelting™ Complete Anesthesia System was designed specifically for stereotaxic surgery. Includes a tabletop and/or mobile cart with Isoflurane Vaporizer, Active Gas Scavenger and continuous flow Induction Chamber.

Inhalant Anesthesia System for Stereotaxic Surgery with Active Gas Waste Removal inlcudes:

  • Tabletop and/or Mobile Anesthesia System Complete with Isoflurane Vaporizer
  • Active Gas Scavenger
  • Continuous Induction Chamber

Features:

  • No flowmeters to adjust. Flip a toggle switch to on to start flow of fresh gas, indicator confirms flow
  • 3 stations for anesthesia with preset flow rate for any station (500cc-5 LPM)
  • Color coded tubing and connector to help user identify which fresh gas supply tubing goes to which station
  • Pressure gauge to let user know when pressure has been released from system for filling of vaporizer.
  • Generates a 50 LPM negative flow that can be attenuated to appropriate levels and divided between 3 stations.
  • Direct vacuum port for use with Vapor-Vac Hooded Induction Chamber
    Adjustable negative flow meters easily activated with toggle switches.
  • Waste anesthetic gases can be discharged into activated charcoal canister or any non re-circulating hood, vent, downdraft/ back draft system or snorkel.
  • Available without negative flow generating pump- can then be used to attenuate active, in-house vacuum source.
  • Designed for use with the Posi-vac Non-Rebreathing system, Vapor-Vac hooded induction chamber, Stereotaxic nose cone or SCOOP, 3EX Vaporizer. (not included)
  • Higher Throughput
  • Intuitive system- easy to operate, precise, safe flow rate for fresh gas
    Healthy for subjects- less morbidity, less mortality

Specifications

For Use With (Equipment) Stereotaxic surgery
Includes Tabletop/Mobile w/Induction Chamber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.