Learn More
Braintree Scientific Gas Anesthetizing Box, 10"" x 8"" x 8""
SDP
Catalog No. NC9807072
Encompass
Gas Anesthetizing Box For use with rodents. The .25" thick transparent plastic anesthetizing box allows for constant observation while anesthetizing. Helps protect against accidental overdosing. For use with rats and mice. Easy to move machine tooled sliding top. Made of durable plastic, this box should last for years. Not for use with liquid organic solvents. **Sold for research purposes only and not for human use**
Catalog No. NC9807072 Supplier Braintree Scientific Supplier No. AB2
Gas Anesthetizing Box For use with rodents. The .25" thick transparent plastic anesthetizing box allows for constant observation while anesthetizing. Helps protect against accidental overdosing. For use with rats and mice. Easy to move machine tooled sliding top. Made of durable plastic, this box should last for years. Not for use with liquid organic solvents. **Sold for research purposes only and not for human use**