Learn More
Sigma Aldrich Fine Chemicals Biosciences Gas bag capacity 1 gal1EA
Catalog No. 502901859
Encompass
for collecting and storing large samples of gases. Used with PVC stopcocks Z186767. Heavy natural rubber.
Catalog No. 50-290-1859 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1867401EA
for collecting and storing large samples of gases. Used with PVC stopcocks Z186767. Heavy natural rubber.